Learn More
CD79b (Igβ) Mouse anti-Human, APC, Clone: SN8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 335800
Description
The CD79b antibody, clone SN8 (3A2-2E7-1F5), is generated from the hybridization of NS-1 mouse myeloma cells with spleen cells from BALB/c mice immunized with cell membrane preparations from B-prolymphocytic leukemia (B-PLL) cells.
The CD79b antibody recognizes an epitope on the extracellular domain of a 36–40 kilodalton (kDa) type I membrane glycoprotein. Immunoglobulin (Ig) antigen receptors are composed of a non-covalently associated complex of Ig and two other proteins, Igα and Igβ, which have been designated as CD79a and CD79b, respectively.

Specifications
CD79b (Igβ) | |
Monoclonal | |
100μg/mL | |
Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
Cell membranes from human B-prol ymphocytic leukemia (B-PLL) cells | |
RUO | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry | |
SN8 | |
APC | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.