Learn More
CD8 Mouse anti-Human, Alexa Fluor 488, Clone: RPA-T8, BD
Mouse Monoclonal Antibody
$235.00 - $373.50
Specifications
Antigen | CD8 |
---|---|
Clone | RPA-T8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | Alexa Fluor 488 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
BDB557704
|
BD Biosciences
557704 |
50 Tests |
Each for $235.00
|
|
|||||
BDB557696
|
BD Biosciences
557696 |
100 Tests |
Each for $373.50
|
|
|||||
Description
Clone RPA-T8 reacts with the α subunit (32kDa) of the two-chain complex expressed on T-cytoxic/suppressor cell populations. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. RPA-T8 reacts with approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. RPA-T8 and HIT8a (Mfr. No.555337) are not cross-blocking.
Host Species: Mouse
Clone: RPA-T8
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
CD8 | |
Flow Cytometry | |
Alexa Fluor 488 | |
RUO | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
Human CD8a | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
RPA-T8 | |
Monoclonal | |
Mouse | |
Immunology | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.