Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8 Mouse anti-Feline, Lion, Tsushima Leopard Cat, R-PE, Clone: fCD8, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 812009
Description
SouthernBiotech offers anti-feline primary antibodies which are suitable flow cytometry and microscopy applications.
Specifications
CD8 | |
Monoclonal | |
0.1mg/mL | |
Supplied in PBS/NaN3 and a stabilizing agent | |
Feline thymocytes | |
RUO | |
Feline, Lion | |
IgG1 |
Flow Cytometry | |
fCD8 | |
R-PE | |
Mouse | |
0.1 mg | |
Primary | |
Store at 2-8°C; Protect from light; Do not freeze |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction