Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BioLegend™ anti-CD8, Clone: SK1
Supplier: BioLegend™ 344714
Description
Host Species: Mouse
Clone: SK1
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
| CD8 | |
| SK1 | |
| APC-Cyanine7 | |
| Unpurified | |
| Primary | |
| IgG1 κ |
| Monoclonal | |
| 0.5mg/mL | |
| Mouse | |
| 100 Tests | |
| Human |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction