Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, FITC, Clone: SK1, BD

Catalog No. BDB347313 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB347313 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB347313 Supplier BD Biosciences Supplier No. 347313
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD8 recognizes the 32-kilodalton (kd) α-subunit of a disulfide-linked bimolecular complex.The majority of peripheral blood CD8+ T lymphocytes expresses an α/β heterodimer (Mr 32, 30 kd), while CD8+ CD16+ natural killer (NK) lymphocytes and CD8+ T-cell receptor (TCR)-γ/δ + T lymphocytes express an α/α homodimer (Mr 30 kd). CD8+ TCR-α/β+ T lymphocytes can express either an α/α homodimer or α/β heterodimer. The CD8 antigenic determinant binds to class I major histocompatibility (MHC) molecules, resulting in increased adhesion between the CD8+ T lymphocytes and target cells.Binding of the CD8 antigen to class I MHC molecules enhances the activation of resting T lymphocytes.The CD8 antigen is coupled to a protein tyrosine kinase, p56 lck. The CD8:p56 lck complex can play a role in T-lymphocyte activation through mediation of the interactions between the CD8 antigen and the CD3/TCR complex. The CD8 antigen is present on the human suppressor/cytotoxic T-lymphocyte subset as well as on a subset of NK lymphocytes.The CD8 antigen is expressed on 19% to 48% of normal peripheral blood lymphocytes and 60% to 85% of normal thymocytes.The CD8+ T- and NK-lymphocyte subsets can be further subdivided into the following groups: CD16+ NK lymphocytes that can express the CD8 antigen in low density;CD57+ T lymphocytes that express high-density CD8 antigen;and CD8+CD62L+ lymphocytes that collaborate with CD8+ CD62Ln++hocytes to generate suppression of B-lymphocyte function.CD8 cross-reacts with lymphocytes of some nonhuman primate species

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone SK1
Concentration 12.5μg/mL
Conjugate FITC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human Peripheral Blood T Cells
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.