Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse CD8 Rat anti-Mouse, Phycoerythrin, Clone: 3.155, MilliporeSigma™

Catalog No. FCMAB428PE Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB428PE 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB428PE Supplier MilliporeSigma Supplier No. FCMAB428PE

Rat Monoclonal Antibody

Milli-Mark™ Anti-mouse CD8-PE Antibody, clone 3.155 is an antibody against mouse CD8 for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone 3.155
Conjugate PE
Formulation Purified rat monoclonal IgMk conjugated to PE in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_001074579
Gene Symbols MAL, p322, CD8, CD8a, Leu2
Host Species Rat
Immunogen Cloned mouse cytolytic T cells.
Purification Method Ion Exchange Purification
Quantity 100 Tests
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.