Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, PE, Clone: RPA-T8, BD

Catalog No. BDB557086 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557086 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557086 Supplier BD Biosciences Supplier No. 557086
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Clone RPA-T8 reacts with the α subunit (32kDa) of the two-chain complex expressed on T-cytoxic/suppressor cell populations. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. RPA-T8 reacts with approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. RPA-T8 and HIT8a (Mfr. No.555337) are not cross-blocking. This clone also reacts with a subset of peripheral blood lymphocytes, but not monocytes nor granuloyctes, of baboon and both rhesus and cynomolgus macaque monkey. In general, a higher frequency of CD8+ and CD4+CD8+ lymphocytes are observed in non-human primates compared to normal human donors. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone RPA-T8
Conjugate PE
Description CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32
Formulation Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD8a
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Human, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.