Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8 Mouse anti-Human, PE-Cy5, Clone: HIT8A, BD

Catalog No. BDB561946 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561946 25 Tests
BDB555636 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB561946 Supplier BD Biosciences Supplier No. 561946

Mouse Monoclonal Antibody

Reacts with the α subunit (32kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 are not cross-blocking. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.

Host Species: Mouse
Clone: HIT8a
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8
Applications Flow Cytometry
Classification Monoclonal
Clone HIT8A
Conjugate PE-Cyanine5
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.