Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Memory CD8 T Cell Enrichment Set - DM anti-Human, BD IMag-DM, Clone: HIT3a, BD

Catalog No. BDB560108 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560108 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560108 Supplier BD Biosciences Supplier No. 560108

Monoclonal Antibody

Cell Separation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Human Memory CD8 T Cell Enrichment Set - DM
Applications Flow Cytometry
Classification Monoclonal
Clone HIT3a
Conjugate BD IMag-DM
Purification Method Affinity Purified
Quantity 5 mL
Regulatory Status RUO
Primary or Secondary Primary
Includes Streptavidin Particles Plus-DM (7.5mL), Human Memory CD8 T Cell Enrichment Cocktail (5mL)
Target Species Human
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.