Learn More
CD8 Mouse anti-Human, V450, Clone: RPA-T8, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560348
Description
Clone RPA-T8 reacts with the α subunit (32kDa) of the two-chain complex expressed on T-cytoxic/suppressor cell populations. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. RPA-T8 reacts with approximately 13-48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. RPA-T8 and HIT8a (Mfr. No. 555337) are not cross-blocking. The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Host Species: Mouse
Clone: RPA-T8
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD8 | |
Monoclonal | |
Violet Fluor 450 (V450) | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
RPA-T8 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human CD8a | |
30 Tests | |
Immunology | |
Baboon, Cynomolgus, Human, Rhesus Monkey | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.