Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 Mouse anti-Rat, Biotin, Clone: 3H5, BD

Catalog No. BDB555013 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555013 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555013 Supplier BD Biosciences Supplier No. 555013

Mouse Monoclonal Antibody

The 3H5 antibody reacts with CD80 (B7-1), a member of the Ig superfamily of transmembrane proteins. CD80, a ligand for CD28 and CD152 (CTLA-4), is one of the accessory molecules that plays an important role in T cell-B cell costimulatory interactions. CD80 is predominantly expressed on antigen-presenting cells. It can be induced on splenic B cells by in vitro polyclonal activation such as LPS treatment. 3H5 mAb is reported to block the costimulatory function of rat CD80 and to immunoprecipitate CD80 from Lewis-S1 cell lysates.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80
Applications Flow Cytometry
Classification Monoclonal
Clone 3H5
Concentration 0.5mg/mL
Conjugate Biotin
Description B7-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat HTLV-1-transformed T-cell line Lewis-S1
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.