Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD80 Mouse anti-Human, Unlabeled, Clone: BB1, BD

Catalog No. BDB555681 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555681 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555681 Supplier BD Biosciences Supplier No. 555681
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 60kDa transmembrane glycoprotein which was clustered as CD80 in the Fifth International Workshop on Human Differentiation Antigens. There have been several recent studies reporting discrepancies in the expression and function of CD80 when the BB1 clone is used compared to other anti-CD80 monoclonals (e.g., L307.4). Caution should therefore be used in the interpretation of data generated using BB1, as reports indicate that BB1 may recognize, in addition to CD80, CD74 and even possibly another as yet undefined molecule. CD80, a member of the Ig supergene family, is expressed on activated B cells, macrophages, and dendritic cells. It is the ligand for two molecules expressed on T cells: CD28 and CD152 (CTLA-4). CD80 is also expressed in activated CD4+ and CD8+ T cells, appearing late after activation, suggesting that activated T cells may be capable of autocrine costimulation via the CD28 activation pathway. The binding of CD28 molecule by CD28 mAb or by CD80 antigen results in T-cell activation and a signal for IL-2 production.

Host Species: Mouse
Clone: BB1
Isotype: IgM, κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD80
Applications Flow Cytometry
Classification Monoclonal
Clone BB1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description B7-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.