Learn More
CD81 Hamster anti-Mouse/Rat, Biotin, Clone: EAT2, BD
Hamster Monoclonal Antibody
Supplier: BD Biosciences 559518
Description
The Eat2 antibody reacts with CD81, a 26kDa nonglycosylated member of the transmembrane 4 integral membrane protein superfamily, expressed by many types of cells. For example, CD81 participates with CD19 and CD21 in the signal transduction complex associated with the B-cell receptor on human B lymphocytes and with the CD4 and CD8 co-receptors on human thymocytes and T lymphocytes. In mouse fetal thymic organ culture, interactions of immature thymocytes with CD81 expressed by thymic stromal cells are required to induce development of T cells with αβ T-cell receptors. Furthermore, CD81 has been shown to play a role in the regulation of rat mastcell degranulation. Despite its important roles in the immune response and wide tissue distribution, CD81-deficient mice are born without obvious developmental abnormalities. However, these mice have abnormal immune responses, and impaired fertility. Eat2 mAb cross-reacts with the rat CD81 antigen. Eat2 mAb was reported cross-reacting with the rat CD81 antigen.
Flow Cytometry

Specifications
CD81 | |
Monoclonal | |
0.5mg/mL | |
TAPA-1; Tspan28; Tapa1; Tapa-1; Target of the antiproliferative antibody 1; | |
Hamster | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
EAT2 | |
Biotin | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
CD81+ mouse B lymphoma 38C13 | |
0.5 mg | |
Immunology | |
Mouse, Rat | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.