Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD81 Hamster anti-Mouse/Rat, PE, Clone: EAT2, BD

Catalog No. BDB559519 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559519 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559519 Supplier BD Biosciences Supplier No. 559519
Add to Cart
Add to Cart

Hamster Monoclonal Antibody

The Eat2 antibody reacts with CD81, a 26kDa nonglycosylated member of the transmembrane 4 integral membrane protein superfamily, expressed by many types of cells. For example, CD81 participates with CD19 and CD21 in the signal transduction complex associated with the B-cell receptor on human B lymphocytes and with the CD4 and CD8 co-receptors on human thymocytes and T lymphocytes. In mouse fetal thymic organ culture, interactions of immature thymocytes with CD81 expressed by thymic stromal cells are required to induce development of T cells with αβ T-cell receptors. Furthermore, CD81 has been shown to play a role in the regulation of rat mastcell degranulation. Despite its important roles in the immune response and wide tissue distribution, CD81-deficient mice are born without obvious developmental abnormalities. However, these mice have abnormal immune responses, and impaired fertility. Eat2 mAb cross-reacts with the rat CD81 antigen. Eat2 mAb was reported cross-reacting with the rat CD81 antigen.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD81
Applications Flow Cytometry
Classification Monoclonal
Clone EAT2
Concentration 0.2mg/mL
Conjugate PE
Description TAPA-1; Tspan28; Tapa1; Tapa-1; Target of the antiproliferative antibody 1;
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Hamster
Immunogen CD81+ mouse B lymphoma 38C13
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.