Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD83 Rat anti-Mouse, APC, Clone: MICHEL-19, BD

Catalog No. BDB558206 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558206 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558206 Supplier BD Biosciences Supplier No. 558206
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The Michel-19 antibody reacts with CD83, a member of the immunoglobulin superfamily that is expressed on mature dendritic cells and activated T lymphocytes. Furthermore, thymic cortical epithelial cells express Cd83 transcripts. CD83 is involved in the regulation of T-cell development and immune responses, and its ligand is found on a subpopulation of splenic B lymphocytes.

Host Species: Rat
Clone: Michel-19
Isotype: IgG1 κ
Species Reactivity: Mouse
Immunogen: Mouse CD83 Recombinant Protein

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD83
Applications Flow Cytometry
Classification Monoclonal
Clone MICHEL-19
Concentration 0.2mg/mL
Conjugate APC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CD83 Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.