Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD83 Rat anti-Mouse, Alexa Fluor 488, Clone: Michel-19, BD

Catalog No. BDB563539 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB563539 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB563539 Supplier BD Biosciences Supplier No. 563539
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The Michel-19 monoclonal antibody specifically binds to CD83. CD83 is a type 1 transmembrane glycoprotein and member of the immunoglobulin superfamily. It is expressed on mature dendritic cells and activated T lymphocytes. Furthermore, thymic cortical epithelial cells express Cd83 transcripts. CD83 is involved in the regulation of T-cell development and immune responses, and its ligand is found on a subpopulation of splenic B lymphocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD83
Applications Flow Cytometry
Classification Monoclonal
Clone Michel-19
Concentration 0.2mg/mL
Conjugate Alexa Fluor 488
Description Cd83; CD83 antigen
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse CD83 Recombinant Protein
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.