Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD85j Mouse anti-Human, Unlabeled, Clone: GHI/75, BD

Catalog No. BDB555941 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555941 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555941 Supplier BD Biosciences Supplier No. 555941
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD85 molecules belong to a large immunoregulatory family and it has been clustered into different subclasses from CD85a to CD85m in the VIIth HLDA workshop. CD85j is also called as Ig-like transcript (ILT2), or leukocyte Ig-like receptor (LIR-1). Reacts with an 110kDa membrane glycoprotein expressed on a subset of NK cells, which varies amongst individuals, and a subpopulation of T lymphocytes. Expression on T lymphocytes, NK cells may depend on the individuals tested. Function studies show that ligation of ILT2 with MHC class I
including HLA-A, B, G1 and -E induces an inhibitory signal via recruitment of SHP-1 phosphatase. This antibody is routinely tested by flow cytometric analysis and fluorescence microscopy. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: GHI/75
Isotype: IgG2b, κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD85j
Applications Flow Cytometry
Classification Monoclonal
Clone GHI/75
Concentration 0.5mg/mL
Conjugate Unconjugated
Description CD85
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.