Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD86 (B7-2) Rat anti-Mouse, FITC, Clone: GL-1 (GL1), MilliporeSigma™
Rat Monoclonal Antibody
Supplier: MilliporeSigma MABF457
Description
This rat monoclonal antibody Anti-CD86 (B7-2) (mouse), FITC, clone GL-1 validated for use in immunohistochemistry, Immunofluorescence and Flow Cytometry for the detection of CD86 (B7-2).
Specifications
CD86 (B7-2) | |
Monoclonal | |
0.5mg/mL | |
Purified Rat Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide. | |
Rat | |
Size Exclusion Chromatography | |
Primary | |
2-8°C, protect from prolonged exposure to light. Do not Freeze. | |
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
GL-1 (GL1) | |
FITC | |
Cd86 | |
Proprietary immunogen | |
100 μg | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction