Learn More
CD86 Mouse anti-Human, Biotin, Clone: IT2.2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555664
Description
IT2.2 recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells including lymphoid cells in germinal centers and Epstein-Barr virus transformed B-cell lines. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in costimulation of T cells in primary immune response. Competitive binding assays demonstrate that, while both IT2.2 and FUN-1 (anti-CD86) recognize the same molecule, they react with different epitopes. IT2.2 blocks costimulation activity of CD86 in functional studies and blocks binding of human CTLA-4-Ig fusion protein to B70 gene-transfected cells.
Host Species: Mouse
Clone: IT2.2
Isotype: IgG2b, κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD86 | |
Monoclonal | |
Biotin | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
IT2.2 | |
B70/B7-2 | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.