Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD86, Clone: 1F8, Abnova™

Catalog No. 89300234 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-234 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-234 Supplier Abnova Corporation Supplier No. H00000942M07
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen CD86
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CD86.
Formulation PBS with no preservative; pH 7.4
Gene CD86
Gene Accession No. BC040261.1
Gene Alias B7-2/B70/CD28LG2/LAB72/MGC34413
Gene Symbols CD86
Host Species Mouse
Immunogen CD86 (AAH40261.1, 24 a.a. ∼ 246 a.a) partial recombinant protein.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.