Learn More
CD86 Mouse anti-Human, Unlabeled, Clone: 2331 (FUN-1), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555655
Description
2331 (FUN-1) recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86) recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. 2331 (FUN-1) blocks costimulation activity of CD86 in functional studies. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: 2331 (FUN-1)
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD86 | |
Monoclonal | |
0.5mg/mL | |
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
2331 (FUN-1) | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Human HBL-1 Cell Line | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.