Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD86 molecule, Mouse, Clone: 3D7, Abnova™

Catalog No. 89014071 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-071 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-071 Supplier Abnova Corporation Supplier No. H00000942M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant CD86.

This gene encodes a type I membrane protein that is a member of the immunoglobulin superfamily. This protein is expressed by antigen-presenting cells, and it is the ligand for two proteins at the cell surface of T cells, CD28 antigen and cytotoxic T-lymphocyte-associated protein 4. Binding of this protein with CD28 antigen is a costimulatory signal for activation of the T-cell. Binding of this protein with cytotoxic T-lymphocyte-associated protein 4 negatively regulates T-cell activation and diminishes the immune response. Alternative splicing results in two transcript variants encoding different isoforms. Additional transcript variants have been described, but their full-length sequences have not been determined. [provided by RefSeq

Sequence: LETDKTRLLSSPFSIELEDPQPPPDHIPWITAVLPTVIICVMVFCLILWKWKKKKRPRNSYKCGTNTMEREESEQTKKREKIHIPERSDETQRVFKSSKTSSCDKSDTCF

Specifications

Antigen CD86
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 3D7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant CD86.
Formulation PBS with no preservative; pH 7.4
Gene CD86
Gene Accession No. BC040261
Gene Alias B7-2/B70/CD28LG2/LAB72/MGC34413
Gene Symbols CD86
Host Species Mouse
Immunogen CD86 (AAH40261, 220 a.a. ∼ 329 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 942
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.