Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD86 Mouse anti-Human, Unlabeled, Clone: IT2.2, BD

Catalog No. BDB555663 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555663 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555663 Supplier BD Biosciences Supplier No. 555663
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IT2.2 recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells including lymphoid cells in germinal centers and Epstein-Barr virus transformed B-cell lines. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in costimulation of T cells in primary immune response. Competitive binding assays demonstrate that, while both IT2.2 and FUN-1 (anti-CD86) recognize the same molecule, they react with different epitopes. IT2.2 blocks costimulation activity of CD86 in functional studies and blocks binding of human CTLA-4-Ig fusion protein to B70 gene-transfected cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD86
Applications Flow Cytometry
Classification Monoclonal
Clone IT2.2
Concentration 0.5mg/mL
Conjugate Unconjugated
Description B70/B7-2
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.