Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD86 Mouse anti-Human, FITC, Clone: 2331 (FUN-1), BD

Catalog No. BDB557343 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB557343 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB557343 Supplier BD Biosciences Supplier No. 557343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

2331 (FUN-1) recognizes a 75kDa cell surface protein, CD86 (B70/B7-2), expressed primarily on monocytes and activated B cells. Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86) recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. 2331 (FUN-1) blocks costimulation activity of CD86 in functional studies. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD86
Applications Flow Cytometry
Classification Monoclonal
Clone 2331 (FUN-1)
Conjugate FITC
Description B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HBL-1 Cell Line
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Cynomolgus, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.