Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD86 Mouse anti-Human, FITC, Clone: 2331 (FUN-1), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 560958
Description
The 2331 (FUN-1) antibody blocks the costimulatory activity of CD86 when tested in functional studies. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Applications: Flow Cytometry

Specifications
CD86 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human HBL-1 Cell Line | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
2331 (FUN-1) | |
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction