Learn More
CD86 Mouse anti-Human, PE-Cy5, Clone: 2331 (FUN-1), BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 555659
Description
Competitive binding assays demonstrate that, while both 2331 (FUN-1) and IT2.2 (anti-CD86) recognize the same molecule, they react with different epitopes. CD86 is the second ligand for CD28 and CTLA-4 and may play an important role in co-stimulation of T cells in primary immune response. 2331 (FUN-1) blocks costimulation activity of CD86 in functional studies. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Host Species: Mouse
- Clone: 2331 (FUN-1)
- Isotype: IgG1 κ
- Species Reactivity: Human
- Applications: Flow Cytometry

Specifications
CD86 | |
Monoclonal | |
PE-Cyanine5 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human HBL-1 Cell Line | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
2331 (FUN-1) | |
B7.2; B7-2; B-lymphocyte activation antigen B7-2; B70; BU63; CD28LG2; LAB72 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.