Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD87 Mouse anti-Human, Unlabeled, Clone: VIM5, BD

Catalog No. BDB555767 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555767 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555767 Supplier BD Biosciences Supplier No. 555767
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with the urokinase plasminogen activator receptor (uPA-R), a 50-65kDa single chain glycosylphosphatidylinositol (GPI)-linked glycoprotein. uPA-R was clustered as CD87 in the Fifth Human Leukocyte Differentiation Antigens Workshop. It is expressed on neutrophils, monocytes, tissue macrophages, endothelial cells, and activated T cells. CD87 plays a role in cell-associated fibrinolytic activity and tissue invasion. This antibody may be useful in the study of myeloid cell differentiation. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: VIM5
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD87
Applications Flow Cytometry
Classification Monoclonal
Clone VIM5
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.