Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD88 Rabbit anti-Human, Unlabeled, Clone: C85-2506, BD

Catalog No. BDB550733 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550733 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550733 Supplier BD Biosciences Supplier No. 550733
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

The human C5a receptor (C5aR) is expressed on granulocytes, monocytes, macrophages mast cells human cell lines of myeloid origin, and certain cell types in several organs, including dendritic cells of secondary lymphoid organs, liver hepatocytes, lung bro

Host Species: Rabbit
Clone: C85-2506
Isotype: IgG
Species Reactivity: Human

Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD88
Applications Immunohistochemistry
Classification Monoclonal
Clone C85-2506
Conjugate Unconjugated
Description C5a Receptor
Formulation Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide.
Host Species Rabbit
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.