Learn More
BioLegend™ anti-CD8a, Clone: 53-6.7
Description
Host Species: Rat
Clone: 53-6.7
Isotype: IgG2a, κ
Cross Reactivity: Mouse
Immunogen: Mouse thymus or spleen
Flow Cytometry
Specifications
Specifications
| Antigen | CD8a |
| Classification | Monoclonal |
| Clone | 53-6.7 |
| Concentration | 0.2mg/mL |
| Conjugate | APC-Cyanine7 |
| Host Species | Rat |
| Immunogen | Mouse thymus or spleen |
| Purification Method | Unpurified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.