Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8α Mouse anti-Porcine, Unconjugated, Clone: 76-2-11, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 452001
Description
SouthernBiotech anti-porcine primary antibodies are available in a variety of conjugated formats including Alexa Fluor.
Specifications
CD8α | |
Monoclonal | |
0.5mg/mL | |
Supplied in borate buffered saline, pH 8.2; No preservatives or amine-containing buffer salts added | |
Fresh dd miniature swine thymocytes | |
RUO | |
Pig | |
Liquid |
Blocking Assay, Depletion Assay, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation | |
76-2-11 | |
Unconjugated | |
Mouse | |
0.5 mg | |
Primary | |
Store at 2-8°C | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction