Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD8α Mouse anti-Chicken, FITC, Clone: EP72, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 839002
Description
SouthernBiotech is a leading provider of anti-chicken primary antibodies which are available in numerous formats including Alexa Fluor and Pacific Blue.
Specifications
CD8α | |
Monoclonal | |
0.5mg/mL | |
Supplied in PBS/NaN3 | |
Chicken splenocytes | |
RUO | |
Chicken | |
IgG2b |
Flow Cytometry, Immunohistochemistry (Frozen) | |
EP72 | |
FITC | |
Mouse | |
0.5 mg | |
Primary | |
Store at 2-8°C; Protect from light |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title