Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-CD8a PE, Clone: 53-6.7,

Catalog No. MABF1544MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF1544MI 100 μg
MABF1543MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABF1544MI Supplier MilliporeSigma™ Supplier No. MABF1544
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Specifically detects CD8a Clone: 53-6.7 in Mouse samples, and it is validated for Flow Cytometry

The 53-6.7 antibody reacts with the 32-34kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8a
Applications Flow Cytometry
Classification Monoclonal
Clone 53-6.7
Conjugate PE
Formulation Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Gene Accession No. P01731
Gene Symbols CD8a
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Purified by size-exclusion chromatography
Quantity 100 μg
Regulatory Status RUO
Research Discipline Inflammation, Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.