Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-CD8a PerCP-Cy5.5, Clone: 53-6.7,
Rat Monoclonal Antibody
$189.29 - $324.48
Specifications
Antigen | CD8a |
---|---|
Clone | 53-6.7 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PerCP-Cyanine5.5 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABF1547MI
|
MilliporeSigma™
MABF1547 |
25 μg |
Each for $189.29
|
|
|||||
MABF1548MI
|
MilliporeSigma™
MABF1548 |
100 μg |
Each for $324.48
|
|
|||||
Description
Specifically detects CD8a Clone: 53-6.7 in Mouse samples, and it is validated for Flow Cytometry
The 53-6.7 antibody reacts with the 32-34kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.Specifications
CD8a | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
Rat | |
Inflammation, Immunology | |
Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
CD8a | |
IgG2a κ | |
Purified by size-exclusion chromatography |
53-6.7 | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
P01731 | |
Proprietary immunogen | |
Primary | |
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title