Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD8b Mouse anti-Rat, FITC, Clone: 341, BD

Catalog No. BDB554973 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554973 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554973 Supplier BD Biosciences Supplier No. 554973
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 341 antibody reacts with the β chain of the CD8 antigen on most thymocytes and a subpopulation of mature T lymphocytes (ie, MHC class I-restricted T cells, including most T suppressor/cytotoxic cells). The CD8 α and β chains (CD8a and CD8b, respectively) form a heterodimer on the surface of most thymocytes and thymus-dependent T suppressor/cytotoxic cells, whereas intestinal intraepithelial lymphocytes, many CD8+ T cells of athymic rats, many activated CD4+ T cells, and most NK cells express CD8a without CD8b. It has been suggested that the expression of the CD8a/CD8b heterodimer is restricted to thymus-derived T lymphocytes. CD8 is an antigen co-receptor on the T cell surface which interacts with MHC class I molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Macrophages have also been reported to express CD8 α and β chains, which are involved in signal transduction. The 341 mAb blocks proliferative and cytotoxic in vitro responses of CD8+ effectors to allogeneic cells.

Host Species: Mouse
Clone: 341
Isotype: IgG1 κ
Species Reactivity: Rt
Immunogen: CD8-positive Wistar rat splenic T-cell hybridomas

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD8b
Applications Flow Cytometry
Classification Monoclonal
Clone 341
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen CD8-positive Wistar rat splenic T-cell hybridomas
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.