Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Mouse anti-Human, Unlabeled, Clone: M-L13, BD

Catalog No. BDB555370 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555370 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555370 Supplier BD Biosciences Supplier No. 555370
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 24kDa type III transmembrane protein which is expressed on platelets, pre-B cells, monocytes, endothelia, and epithelia. CD9 belongs to a family of membrane proteins called tetraspanins which transverse the membrane four times. CD9 is weakly expressed on resting mature B cells. M-L13 induces platelet aggregation and activation. This antibody is also suitable for staining acetone-fixed, frozen tissue sections.

Host Species: Mouse
Clone: M-L13
Isotype: IgG1 κ
Species Reactivity: Hu

Flow Cytometry, Immunohistochemistry (Frozen), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD9
Applications Flow Cytometry
Classification Monoclonal
Clone M-L13
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.