Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Mouse anti-Human, Mouse, Rabbit, Clone: MM2/57, MilliporeSigma™

Catalog No. CBL162MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL162MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL162MI Supplier MilliporeSigma Supplier No. CBL162
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-CD9 Antibody, clone MM2/57 detects level of CD9 and has been published and validated for use in flow cytometry, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD9
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone MM2/57
Conjugate Unconjugated
Formulation The monoclonal is presented at a concentration of 100μg/1ml in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. We recommend that each laboratory determine an optimum working titre for use in its particular appli
Gene Accession No. NM_001769.2
Gene Symbols CD9, Tspan-29, MRP-1, MIC3, p24, Tetraspanin-29, BA2, BTCC-1, 5H9, TSPAN29, GIG2, DRAP-27, P24
Host Species Mouse
Immunogen Human platelet glycoprotein
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rabbit
Content And Storage 4°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.