Learn More
CD9 Mouse anti-Rat, Unlabeled, Clone: RPM.7, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 551808
Description
The RPM.7 antibody reacts with CD9, a 26kDa member of the transmembrane 4 superfamily. Rat CD9 mRNA is detected in many adult tissues, including neural and hematopoietic tissue. Using RPM.7 mAb in flow cytometric analysis of rat hematopoietic tissues, CD9 is found on platelets, peripheral B- and T-lymphocyte subsets, and leukocyte subpopulations in the bone marrow. In humans, CD9 has been found to be associated with integrins and other cell-surface receptors, and it is suggested to play a role in signal transduction and possibly in regulating cellular adhesive properties. In mice, CD9 has been demonstrated to participate in T-cell costimulation and induction of apoptosis.
Host Species: Mouse
Clone: RPM.7
Isotype: IgG3 κ
Species Reactivity: Rat
Flow Cytometry, Immunoprecipitation, Western Blotting

Specifications
CD9 | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Not Reported | |
0.1 mg | |
Immunology | |
Rat | |
IgG3 κ |
Flow Cytometry | |
RPM.7 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.