Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD9 Mouse anti-Human, V450, Clone: M-L13, BD

Catalog No. BDB561326 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561326 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561326 Supplier BD Biosciences Supplier No. 561326
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The M-L13 monoclonal antibody specifically binds to a 24 kDa type III transmembrane protein that is expressed on platelets, pre-B cells, monocytes, endothelial and epithelial cells. CD9 belongs to a family of membrane proteins called tetraspanins that transverse the membrane four times. CD9 is weakly expressed on resting mature B cells. M-L13 induces platelet aggregation and activation. This antibody is also suitable for staining acetone-fixed, frozen tissue sections.

The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD9
Applications Flow Cytometry
Classification Monoclonal
Clone M-L13
Conjugate Violet Fluor 450 (V450)
Description CD9 antigen (p24); 5H9; BA2; BTCC-1; DRAP-27; GIG2; MIC3; MRP-1; TSPAN29
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.