Learn More
CD90.1 Mouse anti-Rat/Mouse, Unlabeled, Clone: HIS51, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 554892
Description
The HIS51 antibody reacts with the rat Thy-1 antigen (CD90) expressed by hematopoietic stem cells, early myeloid and erythroid cells, immature B lymphocytes in the bone marrow and peripheral lymphoid organs, thymocytes, recent thymic emigrants (a subset of CD45RCperipheral T cells), neurons, glomerular mesangial cells, endothelium at inflammatory sites, mast cells, and bone marrow-derived dendritic cells. Rat dendritic epidermal T cells (DEC) are Thy-1-negative. CD90 is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. In addition, there is evidence in the mouse that CD90 mediates adhesion of thymocytes to thymic stroma. HIS51 antibody cross-reacts with the mouse Thy-1.1 alloantigen of the AKR/J and PL strains, but not Thy-1.2 found on most strains. In the mouse, CD90 is found on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), hematopoietic stem cells, and neurons, but not B lymphocytes.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)

Specifications
CD90.1 | |
Monoclonal | |
0.5mg/mL | |
Thy-1/Thy-1.1 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Flow Cytometry | |
HIS51 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat bone marrow cells | |
0.5 mg | |
Immunology | |
Mouse, Rat | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.