Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90.1 Mouse anti-Rat/Mouse, FITC, Clone: HIS51, BD

Catalog No. BDB554894 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554894 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554894 Supplier BD Biosciences Supplier No. 554894
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The HIS51 antibody reacts with the rat Thy-1 antigen (CD90) expressed by hematopoietic stem cells, early myeloid and erythroid cells, immature B lymphocytes in the bone marrow and peripheral lymphoid organs, thymocytes, recent thymic emigrants (a subset of CD45RCperipheral T cells), neurons, glomerular mesangial cells, endothelium at inflammatory sites, mast cells, and bone marrow-derived dendritic cells. Rat dendritic epidermal T cells (DEC) are Thy-1-negative. CD90 is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. In addition, there is evidence in the mouse that CD90 mediates adhesion of thymocytes to thymic stroma. HIS51 antibody cross-reacts with the mouse Thy-1.1 alloantigen of the AKR/J and PL strains, but not Thy-1.2 found on most strains. In the mouse, CD90 is found on thymocytes, most peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), hematopoietic stem cells, and neurons, but not B lymphocytes.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90.1
Applications Flow Cytometry
Classification Monoclonal
Clone HIS51
Concentration 0.5mg/mL
Conjugate FITC
Description Thy-1/Thy-1.1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Rat bone marrow cells
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.