Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90 Mouse anti-Human, Biotin, Clone: 5E10, BD

Catalog No. BDB555594 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555594 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555594 Supplier BD Biosciences Supplier No. 555594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 25-35kDa molecule expressed on 1-4% of human fetal liver cells, cord blood cells, and bone marrow cells. Anti-CD90 reacts with a subset of immature, CD34+ celsl and a distinct subset of mature CD34- cells that are CD3+ CD4+ . The CD90+ CD34+ population is highly enriched for cells capable of long-term culture. Anti-CD90 is useful for enriching high proliferative potential colony-forming cells (HIPP-CFC) which are primative progenitor cells. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: 5E10
Isotype: IgG1 κ
Species Reactivity: Hu

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90
Applications Flow Cytometry
Classification Monoclonal
Clone 5.00E+10
Concentration 0.5mg/mL
Conjugate Biotin
Description THY1; Thy-1 antigen; Thy-1 membrane glycoprotein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human HEL Cell Line
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Baboon, Canine, Cynomolgus, Human, Pig, Rhesus Monkey
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.