Learn More
Description
Host Species: Mouse
Clone: OX-7
Isotype: IgG1, κ
Species Reactivity: Ms, Rt
Immunogen: Rat Thymocyte Thy-1 Antigen
Flow cytometry
Specifications
Specifications
| Antigen | CD90/CD90.1 |
| Classification | Monoclonal |
| Clone | OX-7 |
| Concentration | 0.2mg/mL |
| Host Species | Mouse |
| Immunogen | Rat Thymocyte Thy-1 Antigen |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.