Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD90 Rat anti-Mouse, Unlabeled, Clone: G7, BD

Catalog No. BDB553016 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB553016 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB553016 Supplier BD Biosciences Supplier No. 553016
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The G7 antibody reacts with the Thy-1.1 and Thy-1.2 alloantigens (CD90.1 and CD90.2, respectively) on thymocytes, peripheral T lymphocytes, some intraepithelial T lymphocytes (IEL, DEC), and neurons, but not B lymphocytes, of all mouse strains. CD90 is a GPI-anchored membrane glycoprotein of the Ig superfamily which is involved in signal transduction. In addition, there is evidence that CD90 mediates adhesion of thymocytes to thymic stroma. Soluble G7 mAb stimulates murine T-cell proliferation and IL-2 secretion, via signalling through the TCR/CD3 complex. mAb G7 has been reported to promote apoptosis of thymocytes and CTL clones, as determined by morphological and biochemical criteria. The Thy-1-triggered thymocyte death was found to be developmentally regulated and targeted mainly within the CD4+CD8+CD3- and CD4+CD8+CD3lo cultured immature thymocytes. G7 antibody does not react with Thy-1 (CD90) on rat thymocytes.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD90
Applications Flow Cytometry
Classification Monoclonal
Clone G7
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Thy-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Rat
Immunogen Mouse T-cell hybridoma C 6/68
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2c κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.