Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CD90/Thy1, APC, Clone: 778053, R&D Systems™

Catalog No. FAB7335A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB7335A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB7335A Supplier R&D Systems Supplier No. FAB7335A
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen CD90/Thy1
Applications Flow Cytometry
Classification Monoclonal
Clone 778053
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P01831
Gene Alias CD90, CD90 antigen, CDw90, FLJ33325, Thy1, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Gene Symbols THY1
Host Species Rat
Immunogen Chinese hamster ovary cell line CHO-derived recombinant mouse CD90/Thy1 Gln20-Lys130 Accession # P01831
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7070
Test Specificity Detects mouse CD90/Thy1 in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD90/Thy1 is observed.
Target Species Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.