Learn More
CD91 Mouse anti-Human, FITC, Clone: A2MR-A2, BD
Mouse Monoclonal Antibody
Supplier: BD Biosciences 550496
Description
Reacts with a 600kDa, type I membrane single protein, also known as the α2 Macroglobulin (α2M) receptor/low density lipoprotein (LDL) receptor. The single chain receptor undergoes cleavage, shortly after synthesis, into the 85kDa transmembrane β chain that non-covalently binds to the extracellular 500-515kDa a chain. It has a broad cellular distribution, but in the hematopoietic system it is expressed on monocyte lineage cells. The α2M/LDL receptor mediates endocytosis of a variety of ligands including α2M-proteinase complexes, plasminogen activators in complex with plasminogen activator inhibitor, Pseudomonas Exotoxin A. Ligand binding to α2M/LDL receptor is followed by rapid transport of the ligand to lysosomes for degradation.
Host Species: Mouse
Clone: A2MR-α2
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry

Specifications
CD91 | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
A2MR-A2 | |
Alpha-2-macroglobulin receptor; α2MR; LRP-1; LDL Receptor-Related Protein-1 | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.