Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD95 Mouse anti-Human, Unlabeled, Clone: EOS9.1, BD

Catalog No. BDB550042 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550042 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550042 Supplier BD Biosciences Supplier No. 550042
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with a 45kD type I membrane molecule also known as APO-1, or Fas antigen. CD95 belongs to the tumor necrosis factor receptor (TNF-R) family of proteins and is expressed on various cells, including activated T cells and B cells. The interaction of CD95, Fas antigen, with Fas ligand is one of the many mechanisms that lead to apoptosis or programmed cell death. The interaction of CD95 expressing target cells with Fas ligand on cytotoxic T cells induces an apoptotic signal leading to target cell death. It has been reported that apoptosis is the
mechanism used by the immune system in clonal deletion of self-reactive T cells during embryological development.

Host Species: Mouse
Clone: EOS9.1
Isotype: IgM, κ
Species Reactivity: Human

Flow Cytometry, Functional Assay

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD95
Applications Flow Cytometry
Classification Monoclonal
Clone EOS9.1
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Fas/APO-1
Formulation Aqueous buffered solution containing no preservative.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgM κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.