Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD95 Mouse anti-Human, Unlabeled, Clone: G254-274, BD

Catalog No. BDB556370 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB556370 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB556370 Supplier BD Biosciences Supplier No. 556370
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Programmed cell death (apoptosis) may be induced in response to a variety of cytotoxic stimuli, including activation of surface receptors. An area of particular interest has been the induction of apoptosis by the receptor-ligand pair: Fas (CD95) and Fas Ligand (FasL). Fas is an ∽45kDa cell surface protein which belongs to the the TNF (tumor necrosis factor receptor family, and is expressed in various tissue and cells including the thymus, liver, ovary and lung. FasL, a member of the TNF ligand family, is expressed on activated T and NK cells. FasL initiates signaling at the cell surface by aggregation of individual Fas recpetors via binding to the multivalent ligand. Antibodies which selectively activate Fas can be used in vitro to mimic the apoptotic response associated with FasL. Both Fas and FasL are thought to play an important role in the apoptotic processes that take place during T cell development. Clone G254-274 recognizes human Fas. A soluble form of recombinant human Fas (lacking the Fas transmembrane domain) was used as immunogen.

Host Species: Mouse
Clone: G254-274
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human Fas

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD95
Applications Western Blot
Classification Monoclonal
Clone G254-274
Concentration 0.5mg/mL
Conjugate Unconjugated
Description Fas/APO-1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human Fas
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.