Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD95 (Fas/APO-1) Mouse anti-Human, APC, Clone: DX2, BD

Catalog No. BDB340481 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB340481 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB340481 Supplier BD Biosciences Supplier No. 340481
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The CD95, clone DX2, is derived from hybridization of mouse Sp2/0 myeloma cells with spleen cells from C3H/He mice immunized with L cells transfected with CD95 antigen.

The CD95 recognizes a 48-kdalton (kDa) antigen (Fas/APO-1) that is a member of the tumor necrosis factor/nerve growth factor (TNF/NGF) superfamily. It is associated with activated cells. Cross linking of CD95 antigen on activated cells can elicit apoptosis.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD95 (Fas/APO-1)
Applications Flow Cytometry
Classification Monoclonal
Clone DX2
Concentration 50μg/mL
Conjugate APC
Formulation Phosphate buffered saline with gelatin and 0.1% sodium azide.
Host Species Mouse
Immunogen Human CD95-transfected L Cells
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.