Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD95 Mouse anti-Human, PE-Cy7, Clone: DX2, BD

Catalog No. BDB561633 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB561633 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB561633 Supplier BD Biosciences Supplier No. 561633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The DX2 monoclonal antibody specifically binds to the human Fas antigen (also called APO-1). This 45 kDa transmembrane cell surface molecule was designated as CD95 at the Fifth HLDA Workshop. Fas is a member of the TNF-receptor superfamily and is also known as Tumor necrosis factor receptor superfamily member 6 (TNFRSF6). It is expressed on a variety of normal and neoplastic cells including activated T and B lymphocytes and some undifferentiated thymocytes. The Fas/CD95 antigen is a polypeptide that plays a role in the programmed sequence of events leading to cell death, termed apoptosis. The DX2 clone specifically reacts with murine L cells, murine L1210 leukemia cells and murine P815 mastocytoma cells transfected with human Fas cDNA but not with untransfected parental cell lines. Crosslinking CD95 with DX2 antibody delivers an apoptotic signal indicating that DX2 recognizes a functional epitope of the CD95 antigen.

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD95
Applications Flow Cytometry
Classification Monoclonal
Clone DX2
Conjugate PE-Cyanine7
Description APO-1; FAS; TNFRSF6; APT1; ALPS1A; FAS1; FASTM; FASLG receptor
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human CD95-transfected L Cells
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.