Learn More
Mouse CD96 APC-conjugated Antibody, R&D Systems™
Rat Monoclonal Antibody
Supplier:  R&D Systems FAB5690A
Catalog No. FAB5690A
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
CD96 Monoclonal specifically detects CD96 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD96
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
CD96, CD96 antigenMGC22596, CD96 molecule, DKFZp667E2122, T cell-activated increased late expression protein, TACTILE, TACTILEincreased late expression, T-cell surface protein tactile
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
10225
Mouse
Purified
Flow Cytometry
630612
Flow Cytometry 10 uL/10^6 cells
Q3U0X8
CD96
Mouse myeloma cell line NS0-derived recombinant mouse CD96 Val22-Met536 Accession # Q3U0X8
100 Tests
Primary
Detects mouse CD96 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CD96v2 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.